Gene Transcripts

Gene Symbol
Niacr1
Entrez Gene ID
353250
Gene Name
niacin receptor 1
Gene Aliases
Gpr109a, Gpr109b, Hcar2, Hm74, Hm74b, Pumag, rHM74b
Chromosome Location
Chr. 12: 38160464 - 38161546 on Build Rnor_6.0
UniGene ID
Rn.214478
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181476.1 NP_852141.1 1 836
GenBank mRNA AB103062.1 - 1 836

View Full Product Details