Gene Transcripts

Gene Symbol
B3galtl
Entrez Gene ID
689765
Gene Name
beta 1,3-galactosyltransferase-like
Gene Aliases
B3galtl-ps1
Chromosome Location
Chr. 12: 6385830 - 6476010 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769054.1 XP_008767276.1 11, 12 1434

View Full Product Details