Gene Transcripts
- Gene Symbol
- Olfr75-ps1
- Entrez Gene ID
- 258186
- Gene Name
- olfactory receptor 75, pseudogene 1
- Gene Aliases
- MOR135-20, Olfr75, V1
- Chromosome Location
- Chr. 11: 73405450 - 73409858 on Build GRCm38
- UniGene ID
- Mm.173657
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DR065662.1 |
- |
2 |
560 |
|
Chr. 11 - 73405747 - 73409826 [-] |
GenBankmRNA |
CB173622.1 |
- |
2 |
520 |
|
Chr. 11 - 73405895 - 73409787 [-] |
View Full Product Details