Gene Transcripts

Gene Symbol
Snhg1
Entrez Gene ID
83673
Gene Name
small nucleolar RNA host gene 1
Gene Aliases
Chromosome Location
Chr. 19: 8723487 - 8726326 on Build GRCm38
UniGene ID
Mm.292489
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002896.3 8 341
GenBank mRNA AK036616.1 - 2 2102
AK043166.1 - 8 342
AK051045.1 - 3 1858


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BQ178183.1 - 3 603 Chr. 19 - 8724663 - 8726299 [+]
GenBankmRNA AA646653.1 - 7 310 Chr. 19 - 8724152 - 8726312 [+]
GenBankmRNA AV515710.1 - 7 320 Chr. 19 - 8724140 - 8726323 [+]
GenBankmRNA AA259375.1 - 3 223 Chr. 19 - 8725107 - 8726417 [+]
GenBankmRNA AV497308.1 - 7 321 Chr. 19 - 8724155 - 8726331 [+]
GenBankmRNA BF120608.1 - 6 490 Chr. 19 - 8724158 - 8726436 [+]
GenBankmRNA BI456898.1 - 7 324 Chr. 19 - 8724140 - 8726443 [+]
GenBankmRNA BB816775.1 - 4 150 Chr. 19 - 8725011 - 8726441 [+]
GenBankmRNA AV460418.1 - 7 325 Chr. 19 - 8724138 - 8726323 [+]
GenBankmRNA AA433008.1 - 7 325 Chr. 19 - 8724140 - 8726432 [+]
GenBankmRNA EL960934.1 - 7 328 Chr. 19 - 8724140 - 8726443 [+]
GenBankmRNA CB589630.1 - 302 Chr. - - [-]
GenBankmRNA BB640482.1 - 8 338 Chr. 19 - 8723490 - 8726328 [+]
GenBankmRNA AA038088.1 - 7 325 Chr. 19 - 8724146 - 8726105 [+]
GenBankmRNA BE448643.1 - 7 314 Chr. 19 - 8724148 - 8726344 [+]
GenBankmRNA U40654.1 - 1 2486 Chr. - - [-]
GenBankmRNA DV053423.1 - 360 Chr. - - [-]
GenBankmRNA BG091353.1 - 7 285 Chr. 19 - 8724177 - 8726261 [+]
GenBankmRNA AA419715.1 - 7 325 Chr. 19 - 8724140 - 8725832 [+]
GenBankmRNA BU898422.1 - 2 52 Chr. 19 - 8725506 - 8726326 [+]
GenBankmRNA BY012584.1 - 8 337 Chr. 19 - 8723490 - 8726092 [+]
GenBankmRNA BF608049.1 - 365 Chr. - - [-]
GenBankmRNA DV044547.1 - 83 Chr. - - [-]
GenBankmRNA AL118113.1 - 7 334 Chr. 19 - 8724138 - 8726273 [+]
GenBankmRNA BY098836.1 - 5 190 Chr. 19 - 8724673 - 8726359 [+]
GenBankmRNA AL034970.1 - 7 334 Chr. 19 - 8724138 - 8726409 [+]
GenBankmRNA AA795219.1 - 5 187 Chr. 19 - 8724676 - 8726370 [+]
GenBankmRNA AA178616.1 - 7 288 Chr. 19 - 8724174 - 8726395 [+]
GenBankmRNA AV461499.1 - 7 318 Chr. 19 - 8724157 - 8726363 [+]
GenBankmRNA AA037960.1 - 7 314 Chr. 19 - 8724152 - 8726343 [+]
GenBankmRNA CX567991.1 - 7 328 Chr. 19 - 8724140 - 8726443 [+]
GenBankmRNA BB680690.1 - 6 275 Chr. 19 - 8724451 - 8726442 [+]
GenBankmRNA W42022.1 - 320 Chr. - - [-]
GenBankmRNA AV570392.1 - 7 305 Chr. 19 - 8724157 - 8726390 [+]
GenBankmRNA AW990131.1 - 7 313 Chr. 19 - 8724149 - 8726394 [+]
GenBankmRNA BM730535.1 - 7 308 Chr. 19 - 8724154 - 8726421 [+]
GenBankmRNA AV492597.1 - 7 327 Chr. 19 - 8724140 - 8726345 [+]
GenBankmRNA AA200918.1 - 5 196 Chr. 19 - 8724669 - 8726420 [+]
GenBankmRNA AV576327.1 - 2 58 Chr. 19 - 8725500 - 8726440 [+]
GenBankmRNA BY111985.1 - 8 338 Chr. 19 - 8723490 - 8725825 [+]
GenBankmRNA BY391063.1 - 5 185 Chr. 19 - 8724677 - 8726441 [+]
GenBankmRNA BY013100.1 - 8 338 Chr. 19 - 8723490 - 8725832 [+]
GenBankmRNA CO813536.1 - 2 599 Chr. 19 - 8725004 - 8725964 [+]
GenBankmRNA AV487936.1 - 7 292 Chr. 19 - 8724170 - 8726374 [+]
GenBankmRNA AV481080.1 - 7 305 Chr. 19 - 8724157 - 8726376 [+]
GenBankmRNA W42055.1 - 323 Chr. - - [-]
GenBankmRNA AV474095.1 - 2 113 Chr. 19 - 8725444 - 8726443 [+]
GenBankmRNA AV569303.1 - 7 298 Chr. 19 - 8724164 - 8726438 [+]
GenBankmRNA BE849804.1 - 2 74 Chr. 19 - 8725484 - 8726443 [+]
GenBankmRNA AA028518.1 - 6 240 Chr. 19 - 8724435 - 8726342 [+]
GenBankmRNA AA543872.1 - 5 185 Chr. 19 - 8724678 - 8726442 [+]
GenBankmRNA BQ416548.1 - 7 316 Chr. 19 - 8724146 - 8726443 [+]
GenBankmRNA AA590725.1 - 2 74 Chr. 19 - 8725484 - 8726270 [+]
GenBankmRNA BX525735.1 - 7 315 Chr. 19 - 8724146 - 8726398 [+]
GenBankmRNA AV445236.1 - 7 331 Chr. 19 - 8724157 - 8726345 [+]
GenBankmRNA DV065694.1 - 180 Chr. - - [-]
GenBankmRNA AV502115.1 - 3 307 Chr. 19 - 8725029 - 8726002 [+]
GenBankmRNA AA220561.1 - 7 303 Chr. 19 - 8724159 - 8726387 [+]
GenBankmRNA EL962085.1 - 7 375 Chr. 19 - 8724140 - 8726432 [+]
GenBankmRNA BG090562.1 - 7 285 Chr. 19 - 8724177 - 8726313 [+]
GenBankmRNA AI155059.1 - 2 295 Chr. 19 - 8725261 - 8726385 [+]
GenBankmRNA AW985636.1 - 7 324 Chr. 19 - 8724140 - 8726383 [+]
GenBankmRNA AV495678.1 - 3 223 Chr. 19 - 8725108 - 8726443 [+]
GenBankmRNA BX528587.1 - 7 326 Chr. 19 - 8724140 - 8726387 [+]
GenBankmRNA CF896937.1 - 7 314 Chr. 19 - 8724148 - 8726442 [+]
GenBankmRNA AV473637.1 - 4 129 Chr. 19 - 8725029 - 8726443 [+]
GenBankmRNA AV584788.1 - 6 272 Chr. 19 - 8724418 - 8726393 [+]
GenBankmRNA BI068068.1 - 2 254 Chr. 19 - 8725304 - 8726388 [+]
GenBankmRNA AV505845.1 - 7 342 Chr. 19 - 8724147 - 8726323 [+]
GenBankmRNA AV579856.1 - 2 72 Chr. 19 - 8725486 - 8726443 [+]
GenBankmRNA AA462480.1 - 7 325 Chr. 19 - 8724140 - 8726311 [+]
GenBankmRNA BY113223.1 - 8 338 Chr. 19 - 8723490 - 8726081 [+]
GenBankmRNA AV469387.1 - 5 161 Chr. 19 - 8724704 - 8726443 [+]
GenBankmRNA BU898430.1 - 2 52 Chr. 19 - 8725506 - 8726326 [+]
GenBankmRNA AV052352.1 - 1 35 Chr. 19 - 8725781 - 8726442 [+]
GenBankmRNA BF608050.1 - 7 326 Chr. 19 - 8724147 - 8726443 [+]
GenBankmRNA AV481116.1 - 7 304 Chr. 19 - 8724158 - 8726391 [+]
GenBankmRNA BF607556.1 - 7 378 Chr. 19 - 8724140 - 8726443 [+]
GenBankmRNA BP427963.1 - 5 197 Chr. 19 - 8724669 - 8726443 [+]
GenBankmRNA AV501497.1 - 7 305 Chr. 19 - 8724157 - 8726389 [+]
GenBankmRNA BB254596.2 - 8 381 Chr. 19 - 8723487 - 8726326 [+]
GenBankmRNA AA110623.1 - 7 292 Chr. 19 - 8724170 - 8725927 [+]
GenBankmRNA AA760260.1 - 6 258 Chr. 19 - 8724140 - 8726396 [+]

View Full Product Details