Gene Transcripts

Gene Symbol
BCAR4
Entrez Gene ID
400500
Gene Name
breast cancer anti-estrogen resistance 4 (non-protein coding)
Gene Aliases
Chromosome Location
Chr. 16: 11819830 - 11828832 on Build GRCh38
UniGene ID
Hs.24611
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024049.1 4 988
NR_024050.1 3 346
NR_131216.1 2 276
NR_131217.1 3 561
NR_131222.1 3 394
GenBank mRNA BX107175.1 - 3 317
H03167.1 - 2 262
HE601934.1 - 1 130

View Full Product Details