Gene Transcripts

Gene Symbol
ADRA1B
Entrez Gene ID
147
Gene Name
adrenoceptor alpha 1B
Gene Aliases
ADRA1, ALPHA1BAR
Chromosome Location
Chr. 5: 159916733 - 159989205 on Build GRCh38
UniGene ID
Hs.368632
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005265819.2 XP_005265876.1 2 1794
GenBank mRNA BX648936.1 - 1 5684

View Full Product Details