Gene Transcripts
- Gene Symbol
- PCA3
- Entrez Gene ID
- 50652
- Gene Name
- prostate cancer associated 3 (non-protein coding)
- Gene Aliases
- DD3, NCRNA00019, PCAT3
- Chromosome Location
- Chr. 9: 76764436 - 76787569 on Build GRCh38
- UniGene ID
- Hs.663766
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DQ374660.1 |
- |
2 |
264 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF103908.3 |
- |
1 |
1964 |
|
Chr. 9 - 76782136 - 76787569 [+] |
View Full Product Details