Gene Transcripts
- Gene Symbol
- Zfp54
- Entrez Gene ID
- 22712
- Gene Name
- zinc finger protein 54
- Gene Aliases
- 2810034D10Rik, KRAB10, Zfp-54, Zfp76, mszf83
- Chromosome Location
- Chr. 17: 21423227 - 21435640 on Build GRCm38
- UniGene ID
- Mm.12916
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK012854.1 |
- |
1 |
565 |
|
View Full Product Details