Gene Transcripts
- Gene Symbol
- Mbd5
- Entrez Gene ID
- 109241
- Gene Name
- methyl-CpG binding domain protein 5
- Gene Aliases
- 9430004D19Rik, AA536666, AI426407, C030040A15Rik, Gm1630, OTTMUSG00000012483
- Chromosome Location
- Chr. 2: 48949508 - 49317070 on Build GRCm38
- UniGene ID
- Mm.440436
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021136.1 |
- |
|
84 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY728278.1 |
- |
1 |
84 |
|
Chr. 2 - 49318853 - 49319525 [+] |
View Full Product Details