Gene Transcripts
- Gene Symbol
- Zzz3
- Entrez Gene ID
- 108946
- Gene Name
- zinc finger, ZZ domain containing 3
- Gene Aliases
- 3110065C23Rik, 6430567E01Rik, AA408566, AI481057, AW556059, R75514
- Chromosome Location
- Chr. 3: 152394486 - 152462826 on Build GRCm38
- UniGene ID
- Mm.379292
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK132612.1 |
- |
3, 4 |
428 |
|
BC060614.1 |
- |
6, 7 |
2193 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB614155.1 |
- |
3, 4 |
422 |
|
Chr. 3 - 152420011 - 152450690 [+] |
View Full Product Details