Gene Transcripts

Gene Symbol
Rimkla
Entrez Gene ID
194237
Gene Name
ribosomal modification protein rimK-like family member A
Gene Aliases
B930030J24, NAAGS, NAAGS-II, Rimk
Chromosome Location
Chr. 4: 119465279 - 119493153 on Build GRCm38
UniGene ID
Mm.206201
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177572.4 NP_808240.2 2 376
GenBank mRNA AK047164.1 - 2 371
AK163360.1 - 2 367
BC057371.1 - 2 357 6835897
BU611721.1 - 2 406

View Full Product Details