Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172113.1 | NP_001165584.1 | 10 | 1209 | |
NM_001172114.1 | NP_001165585.1 | 9 | 1089 | ||
NM_153792.2 | NP_722487.2 | 10 | 1229 | ||
GenBank mRNA | AK088431.1 | - | 9 | 998 | |
AK131617.1 | - | 10 | 1170 | ||
AK179342.1 | - | 1 | 155 | ||
AK217262.1 | - | 2 | 347 | ||
BC008598.1 | - | 10 | 1132 | 3499933 |