Gene Transcripts
- Gene Symbol
- Wdr59
- Entrez Gene ID
- 319481
- Gene Name
- WD repeat domain 59
- Gene Aliases
- 5430401O09Rik, Cdw12
- Chromosome Location
- Chr. 8: 111448781 - 111522107 on Build GRCm38
- UniGene ID
- Mm.247294
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_176923.4 |
NP_795897.2
|
24, 25 |
2596 |
|
GenBank mRNA |
AK030644.1 |
- |
24, 25 |
2477 |
|
View Full Product Details