Gene Transcripts
- Gene Symbol
- Atp10b
- Entrez Gene ID
- 319767
- Gene Name
- ATPase, class V, type 10B
- Gene Aliases
- 5930426O13Rik, 9030605H24Rik
- Chromosome Location
- Chr. 11: 42939557 - 43262285 on Build GRCm38
- UniGene ID
- Mm.252339
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_176999.3 |
NP_795973.2
|
23 |
4378 |
|
GenBank mRNA |
AK220352.1 |
- |
13 |
2524 |
|
View Full Product Details