Gene Transcripts

Gene Symbol
Vmn2r69
Entrez Gene ID
330581
Gene Name
vomeronasal 2, receptor 69
Gene Aliases
Chromosome Location
Chr. 7: 85406376 - 85415676 on Build GRCm38
UniGene ID
Mm.469909
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105182.1 NP_001098652.1 3 810

View Full Product Details