Gene Transcripts

Gene Symbol
Pomc
Entrez Gene ID
24664
Gene Name
proopiomelanocortin
Gene Aliases
Pomc1, Pomc2, alphaMSH
Chromosome Location
Chr. 6: 28382949 - 28388769 on Build Rnor_6.0
UniGene ID
Rn.108195
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139326.2 NP_647542.1 3 353
GenBank mRNA AF510391.1 - 2 232
BC058443.1 - 3 353 6919050

View Full Product Details