Gene Transcripts

Gene Symbol
Mcpt10
Entrez Gene ID
54269
Gene Name
mast cell protease 10
Gene Aliases
Chromosome Location
Chr. 15: 34622126 - 34625141 on Build Rnor_6.0
UniGene ID
Rn.48655
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017146.1 NP_058842.1 280
GenBank mRNA U67913.1 - 77
X68657.1 - 280

View Full Product Details