Gene Transcripts

Gene Symbol
Vps37a
Entrez Gene ID
290775
Gene Name
vacuolar protein sorting 37 homolog A (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 16: 54860811 - 54899409 on Build Rnor_6.0
UniGene ID
Rn.19
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024867.1 NP_001020038.1 6 912
XM_008771289.1 XP_008769511.1 6 974
GenBank mRNA BC098069.1 - 6 912
FQ232149.1 - 6 966

View Full Product Details