Gene Transcripts

Gene Symbol
Slc7a12
Entrez Gene ID
294881
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 12
Gene Aliases
Chromosome Location
Chr. 2: 88559677 - 88593763 on Build Rnor_6.0
UniGene ID
Rn.34357
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011948.1 NP_001011948.1 356
GenBank mRNA BC078940.1 - 356

Gene Symbol
LOC100910566
Entrez Gene ID
100910566
Gene Name
solute carrier family 7 member 13-like
Gene Aliases
Chromosome Location
Chr. 2: 88668360 - 88669438 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006232118.2 XP_006232180.1 1 383

Gene Symbol
LOC361914
Entrez Gene ID
361914
Gene Name
similar to solute carrier family 7 (cationic amino acid transporter, y+ system), member 12
Gene Aliases
Chromosome Location
Chr. 2: 88453792 - 88553113 on Build Rnor_6.0
UniGene ID
Rn.207166
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017465.1 NP_001017465.1 1 356
XM_006232112.2 XP_006232174.1 1 383
GenBank mRNA BC079349.1 - 1 356

View Full Product Details