Gene Transcripts

Gene Symbol
Pcdh18
Entrez Gene ID
295027
Gene Name
protocadherin 18
Gene Aliases
Chromosome Location
Chr. 2: 138819957 - 138833930 on Build Rnor_6.0
UniGene ID
Rn.61115
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100524.1 NP_001093994.1 1 2286
XM_006232322.2 XP_006232384.1 1 2351
GenBank mRNA BC168716.1 - 1 2286

View Full Product Details