Gene Transcripts

Gene Symbol
Sox15
Entrez Gene ID
363632
Gene Name
SRY (sex determining region Y)-box 15
Gene Aliases
Chromosome Location
Chr. 10: 56260514 - 56263503 on Build Rnor_6.0
UniGene ID
Rn.145250
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108830.1 NP_001102300.1 2 419

View Full Product Details