Gene Transcripts

Gene Symbol
Olr60
Entrez Gene ID
405017
Gene Name
olfactory receptor 60
Gene Aliases
Chromosome Location
Chr. 1: 167892099 - 167893061 on Build Rnor_6.0
UniGene ID
Rn.141919
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000748.1 NP_001000748.1 1 484

View Full Product Details