Gene Transcripts

Gene Symbol
Zfp28
Entrez Gene ID
102548693
Gene Name
zinc finger protein 28
Gene Aliases
Zfp28l
Chromosome Location
Chr. 1: 71167198 - 71182994 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228205.2 XP_006228267.1 8 1618
XM_006228206.2 XP_006228268.1 7 642
XM_006228207.2 XP_006228269.1 6 587
XM_006228208.2 XP_006228270.1 7 1531
XM_008759006.1 XP_008757228.1 7 720
XM_008759007.1 XP_008757229.1 7 657

View Full Product Details