Gene Transcripts

Gene Symbol
Mageb7
Entrez Gene ID
503442
Gene Name
melanoma antigen, family B, 7
Gene Aliases
RGD1563904
Chromosome Location
Chr. X: 54938470 - 54941813 on Build Rnor_6.0
UniGene ID
Rn.218619
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109367.1 NP_001102837.1 1 337
XM_006256984.2 XP_006257046.1 2 553

View Full Product Details