Gene Transcripts

Gene Symbol
Atp6ap2
Entrez Gene ID
302526
Gene Name
ATPase, H+ transporting, lysosomal accessory protein 2
Gene Aliases
Chromosome Location
Chr. X: 11137889 - 11164854 on Build Rnor_6.0
UniGene ID
Rn.12944
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007091.1 NP_001007092.1 9 1071
GenBank mRNA AB188298.1 - 9 1071
BC079339.1 - 1071 7106562

View Full Product Details