Gene Transcripts

Gene Symbol
ALDH1L2
Entrez Gene ID
160428
Gene Name
aldehyde dehydrogenase 1 family member L2
Gene Aliases
mtFDH
Chromosome Location
Chr. 12: 105019784 - 105084609 on Build GRCh38
UniGene ID
Hs.42572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034173.3 NP_001029345.2 9 1190
NR_027752.1 9 1186
XM_011537986.2 XP_011536288.1 7 749
XM_011537988.2 XP_011536290.1 9 1211
XM_011537989.2 XP_011536291.1 9 1211
XM_017018889.1 XP_016874378.1 10 1396
XM_017018890.1 XP_016874379.1 9 1211
GenBank mRNA AK095827.1 - 9 1085
AK300373.1 - 9 1085
BC103934.1 - 9 1085 40008174
BC103935.1 - 9 1081 40008176
CR627287.1 - 5 405

View Full Product Details