Gene Transcripts

Gene Symbol
MTPN
Entrez Gene ID
136319
Gene Name
myotrophin
Gene Aliases
GCDP, V-1
Chromosome Location
Chr. 7: 135926755 - 135977456 on Build GRCh38
UniGene ID
Hs.602015
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145808.3 NP_665807.1 4 3255
GenBank mRNA AK055660.1 - 1 1806
AL110137.1 - 1 634
AL834231.1 - 4 3127
AY611627.1 - 1 453

Gene Symbol
LUZP6
Entrez Gene ID
767558
Gene Name
leucine zipper protein 6
Gene Aliases
MPD6, MTPNUT
Chromosome Location
Chr. 7: 135926755 - 135977456 on Build GRCh38
UniGene ID
Hs.602015
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128619.2 NP_001122091.2 4 3255

View Full Product Details