Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286715.1 | NP_001273644.1 | 19 | 2838 | |
NM_001286717.1 | NP_001273646.1 | 19 | 2718 | ||
NM_015239.2 | NP_056054.2 | 20 | 2783 | ||
XM_005251847.3 | XP_005251904.1 | 20 | 2849 | ||
XM_005251848.2 | XP_005251905.1 | 20 | 3027 | ||
XM_005251849.3 | XP_005251906.1 | 19 | 2773 | ||
XM_011518418.2 | XP_011516720.1 | 20 | 2849 | ||
XM_011518420.2 | XP_011516722.1 | 10 | 6102 | ||
XM_017014544.1 | XP_016870033.1 | 20 | 3394 | ||
XM_017014545.1 | XP_016870034.1 | 20 | 2729 | ||
XM_017014546.1 | XP_016870035.1 | 19 | 2616 | ||
XM_017014547.1 | XP_016870036.1 | 19 | 2773 | ||
XM_017014548.1 | XP_016870037.1 | 19 | 2716 | ||
XM_017014549.1 | XP_016870038.1 | 18 | 2735 | ||
GenBank mRNA | AB028958.2 | - | 20 | 2761 | |
AK001544.1 | - | 21 | 2583 | ||
AK022562.1 | - | 8 | 1426 | ||
AK023280.1 | - | 8 | 1418 | ||
AK024698.1 | - | 4 | 368 | ||
AK295774.1 | - | 19 | 2718 | ||
AK299506.1 | - | 16 | 2278 | ||
AL833359.1 | - | 8 | 1375 | ||
BC060815.1 | - | 20 | 2748 | 30344122 | |
BX648366.1 | - | 8 | 1830 |