Gene Transcripts

Gene Symbol
MKRN2
Entrez Gene ID
23609
Gene Name
makorin ring finger protein 2
Gene Aliases
HSPC070, RNF62
Chromosome Location
Chr. 3: 12557014 - 12583713 on Build GRCh38
UniGene ID
Hs.591666
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271707.1 NP_001258636.1 2 330
NM_014160.4 NP_054879.3 3 459
GenBank mRNA AF302084.1 - 3 373
AK091318.1 - 3 404
AK298463.1 - 2 257
BC001799.2 - 1 168
BC015715.1 - 3 401 3916199
DR157266.1 - 3 459

View Full Product Details