Gene Transcripts

Gene Symbol
NUP35
Entrez Gene ID
129401
Gene Name
nucleoporin 35
Gene Aliases
MP-44, MP44, NP44, NUP53
Chromosome Location
Chr. 2: 183117490 - 183161684 on Build GRCh38
UniGene ID
Hs.180591
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287584.1 NP_001274513.1 8 918
NM_001287585.1 NP_001274514.1 8 938
NM_138285.4 NP_612142.2 7 815
NR_109856.1 6 673
XM_006712254.2 XP_006712317.1 7 780
XM_011510576.2 XP_011508878.1 9 1046
XM_011510577.2 XP_011508879.1 9 1233
XM_017003308.1 XP_016858797.1 7 971
GenBank mRNA AF411516.1 - 7 815
AF514993.1 - 7 772
AK289601.1 - 6 616
AK290760.1 - 7 727
AK298199.1 - 8 848
AK302350.1 - 8 887
BC035597.1 - 7 739
BC047029.1 - 7 802 5541023
BC061698.1 - 7 769 4132440
BI821231.1 - 6 609

View Full Product Details