Gene Transcripts

Gene Symbol
R3HDML
Entrez Gene ID
140902
Gene Name
R3H domain containing like
Gene Aliases
dJ881L22.3
Chromosome Location
Chr. 20: 44337158 - 44350792 on Build GRCh38
UniGene ID
Hs.580807
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178491.3 NP_848586.1 4 563
GenBank mRNA BC107048.2 - 4 563

View Full Product Details