Gene Transcripts

Gene Symbol
OR8H1
Entrez Gene ID
219469
Gene Name
olfactory receptor family 8 subfamily H member 1
Gene Aliases
OR11-180
Chromosome Location
Chr. 11: 56290127 - 56291062 on Build GRCh38
UniGene ID
Hs.553628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005199.1 NP_001005199.1 1 445
GenBank mRNA BC136909.1 - 1 473
BC136910.1 - 1 473 9020901

View Full Product Details