Gene Transcripts

Gene Symbol
MAJIN
Entrez Gene ID
283129
Gene Name
membrane anchored junction protein
Gene Aliases
C11orf85
Chromosome Location
Chr. 11: 64938230 - 64972104 on Build GRCh38
UniGene ID
Hs.567793
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037225.2 NP_001032302.1 8, 9 756
XM_006718504.2 XP_006718567.1 8, 9 867
GenBank mRNA BC106951.2 - 7, 8 551
BC106952.1 - 7, 8 551 40035117

View Full Product Details