Gene Transcripts

Gene Symbol
RPL34
Entrez Gene ID
6164
Gene Name
ribosomal protein L34
Gene Aliases
L34
Chromosome Location
Chr. 4: 108620558 - 108630484 on Build GRCh38
UniGene ID
Hs.438227
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000995.4 NP_000986.2 5 359
NM_001319232.1 NP_001306161.1 6 505
NM_001319234.1 NP_001306163.1 5 489
NM_001319235.1 NP_001306164.1 5 439
NM_001319236.1 NP_001306165.1 5 359
NM_033625.3 NP_296374.1 5 439
GenBank mRNA AA128058.1 - 6 464
AF147334.1 - 4 263
AY762981.1 - 2 142
BC001773.1 - 5 373 3355603
BC070208.1 - 5 340 6724916
BC106009.1 - 5 351 4390762
BE394007.1 - 5 405
BU588998.1 - 5 439
CB132938.1 - 5 490
CD175155.1 - 5 461
CD249923.1 - 5 397
CR542232.1 - 4 307
CR542242.1 - 4 307
DA779124.1 - 5 340

View Full Product Details