Gene Transcripts

Gene Symbol
Eif2ak3
Entrez Gene ID
13666
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 3
Gene Aliases
AI427929, Pek, Perk
Chromosome Location
Chr. 6: 70844484 - 70905240 on Build GRCm38
UniGene ID
Mm.247167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010121.2 NP_034251.2 10, 11 1955
GenBank mRNA AF076681.1 - 10, 11 1947
AK139679.1 - 8, 9 1277
BC054809.1 - 10, 11 1967 6839221

View Full Product Details