Gene Transcripts

Gene Symbol
Zbtb7a
Entrez Gene ID
16969
Gene Name
zinc finger and BTB domain containing 7a
Gene Aliases
9030619K07Rik, 9130006G12Rik, AI452336, FBI-1, Lrf, Pokemon, Zbtb7
Chromosome Location
Chr. 10: 81134585 - 81152222 on Build GRCm38
UniGene ID
Mm.20920
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010731.3 NP_034861.3 2 660
GenBank mRNA AF086830.1 - 2 277
AK149961.1 - 2 337
AK154907.1 - 2 354
AK157664.1 - 354
BC024815.2 - 2 660
BC057204.1 - 2 298
BC138524.1 - 2 419 8861544
BC145311.1 - 2 459 9053866
BC171957.1 - 2 419 9054243

View Full Product Details