Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001110796.2 | NP_001104266.1 | 5 | 7950 | |
NM_011995.4 | NP_036125.4 | 5 | 7950 | ||
GenBank mRNA | AB083477.1 | - | 6 | 7839 | |
AB083478.1 | - | 6 | 7839 | ||
Y19185.2 | - | 6 | 7860 | ||
Y19186.2 | - | 6 | 7860 |