Gene Transcripts

Gene Symbol
Thap7
Entrez Gene ID
69009
Gene Name
THAP domain containing 7
Gene Aliases
1810004B07Rik
Chromosome Location
Chr. 16: 17527558 - 17531512 on Build GRCm38
UniGene ID
Mm.10174
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026909.2 NP_081185.1 2, 3 115
GenBank mRNA AK154641.1 - 2, 3 201
BC018221.1 - 2, 3 115 4215606

View Full Product Details