Gene Transcripts

Gene Symbol
1500009L16Rik
Entrez Gene ID
69784
Gene Name
RIKEN cDNA 1500009L16 gene
Gene Aliases
2700050C19Rik, OCC-1
Chromosome Location
Chr. 10: 83722666 - 83762762 on Build GRCm38
UniGene ID
Mm.271188
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145198.1 NP_001138670.1 3 207
GenBank mRNA AK005183.1 - 3 138
BC048550.1 - 3 207

View Full Product Details