Gene Transcripts

Gene Symbol
Lysmd2
Entrez Gene ID
70082
Gene Name
LysM, putative peptidoglycan-binding, domain containing 2
Gene Aliases
2210402C18Rik, AV033872, AW538442
Chromosome Location
Chr. 9: 75625732 - 75637773 on Build GRCm38
UniGene ID
Mm.19119
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027309.2 NP_081585.2 2 461
GenBank mRNA BC048545.1 - 2 450
DT905233.1 - 2 216

View Full Product Details