Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027412.3 | NP_081688.2 | 2 | 509 | |
GenBank mRNA | AK008753.1 | - | 1 | 840 | |
AK018145.1 | - | 2 | 549 | ||
AK169288.1 | - | 2 | 510 | ||
AK169296.1 | - | 2 | 510 | ||
BC035304.1 | - | 1 | 420 | ||
BC043061.1 | - | 1 | 821 | 5704924 | |
BC058967.1 | - | 1 | 821 | 5704924 | |
BC063773.1 | - | 1 | 475 | 30147949 |