Gene Transcripts

Gene Symbol
Tmem87b
Entrez Gene ID
72477
Gene Name
transmembrane protein 87B
Gene Aliases
2610301K12Rik, 2810431I02Rik, AU014804
Chromosome Location
Chr. 2: 128818072 - 128854261 on Build GRCm38
UniGene ID
Mm.259688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028248.2 NP_082524.2 5 504
GenBank mRNA AK011959.1 - 5 425
AK050203.1 - 5 496
AK164504.1 - 5 486
AK166991.1 - 5 484
AK171811.1 - 6 420
AK172611.1 - 6 420
BC031488.1 - 5 460

View Full Product Details