Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252326.1 | NP_001239255.1 | 23 | 3415 | |
NM_001252327.1 | NP_001239256.1 | 23 | 3361 | ||
NM_133992.3 | NP_598753.1 | 23 | 3442 | ||
GenBank mRNA | AK004773.2 | - | 22 | 3441 | |
AK047887.1 | - | 23 | 3402 | ||
AK089001.1 | - | 23 | 3443 | ||
AK129196.1 | - | 21 | 3912 | ||
AK157800.1 | - | 23 | 3319 | ||
AK160655.1 | - | 5 | 564 | ||
BC006064.1 | - | 3 | 398 | ||
BC008521.1 | - | 22 | 3732 | ||
BC043659.1 | - | 10 | 1354 | ||
BC075686.1 | - | 23 | 3419 | 30615204 | |
BC079841.1 | - | 23 | 3376 | 6417806 |