Gene Transcripts

Gene Symbol
Rnpepl1
Entrez Gene ID
108657
Gene Name
arginyl aminopeptidase (aminopeptidase B)-like 1
Gene Aliases
1110014H17Rik, AI894167
Chromosome Location
Chr. 1: 92910825 - 92920585 on Build GRCm38
UniGene ID
Mm.200971
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181405.4 NP_852070.3 4 1049
GenBank mRNA AK154881.1 - 4 1050
AK183521.1 - 2 131
BC031789.1 - 4 469 4006916
BQ559332.1 - 2 233
CA315475.1 - 4 743

View Full Product Details