Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145725.1 | NP_001139197.1 | 6, 7 | 569 | |
NM_017816.2 | NP_060286.1 | 6, 7 | 667 | ||
XM_011513505.1 | XP_011511807.1 | 6, 7 | 969 | ||
XM_011513506.2 | XP_011511808.1 | 5, 6 | 600 | ||
GenBank mRNA | AK000432.1 | - | 6, 7 | 614 | |
AK290750.1 | - | 6, 7 | 644 | ||
AL136750.1 | - | 6, 7 | 569 | ||
BC015796.2 | - | 6, 7 | 629 | 3911275 | |
CR533548.1 | - | 4, 5 | 426 |