Gene Transcripts

Gene Symbol
SNX17
Entrez Gene ID
9784
Gene Name
sorting nexin 17
Gene Aliases
Chromosome Location
Chr. 2: 27370496 - 27377533 on Build GRCh38
UniGene ID
Hs.278569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267059.1 NP_001253988.1 3 433
NM_001267060.1 NP_001253989.1 2 394
NM_001267061.1 NP_001253990.1 3 304
NM_014748.3 NP_055563.1 3 469
NR_049782.1 3 594
NR_049783.1 3 493
NR_049784.1 3 469
GenBank mRNA AK129689.1 - 3 353
AK222543.1 - 3 352
AK293252.1 - 3 369
AK293464.1 - 3 285
AK297054.1 - 3 478
AK298620.1 - 3 304
AK298869.1 - 2 255
AK300144.1 - 3 316
AK316373.1 - 2 174
BC002524.1 - 3 330 3140610
BC002610.1 - 3 319 3143104
BC014620.2 - 3 330 3010648
BC050590.1 - 3 324 6044270
BT007167.1 - 3 221
CR457081.1 - 3 221
D31764.1 - 3 443
DB177194.1 - 3 459

View Full Product Details