Gene Transcripts

Gene Symbol
Nelfa
Entrez Gene ID
305455
Gene Name
negative elongation factor complex member A
Gene Aliases
Whsc2
Chromosome Location
Chr. 14: 82093837 - 82117913 on Build Rnor_6.0
UniGene ID
Rn.9138
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008339.1 NP_001008340.1 9, 10 1374
GenBank mRNA BC085948.1 - 9, 10 1374

View Full Product Details