Gene Transcripts

Gene Symbol
TCOF1
Entrez Gene ID
6949
Gene Name
treacle ribosome biogenesis factor 1
Gene Aliases
MFD1, TCS, TCS1, treacle
Chromosome Location
Chr. 5: 150357639 - 150400308 on Build GRCh38
UniGene ID
Hs.519672
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000356.3 NP_000347.2 7 761
NM_001008657.2 NP_001008657.1 8 992
NM_001135243.1 NP_001128715.1 8 992
NM_001135244.1 NP_001128716.1 8 992
NM_001135245.1 NP_001128717.1 7 761
NM_001195141.1 NP_001182070.1 8 992
XM_005268502.3 XP_005268559.1 8 929
XM_005268503.3 XP_005268560.1 8 929
XM_005268504.3 XP_005268561.1 8 929
XM_005268505.3 XP_005268562.1 8 929
XM_005268506.3 XP_005268563.1 8 929
XM_005268507.3 XP_005268564.1 7 698
XM_005268508.3 XP_005268565.1 8 929
XM_005268509.3 XP_005268566.1 8 929
XM_011537678.2 XP_011535980.1 8 929
XM_017009792.1 XP_016865281.1 8 929
XM_017009793.1 XP_016865282.1 8 929
XM_017009794.1 XP_016865283.1 8 929
XM_017009795.1 XP_016865284.1 7 698
GenBank mRNA AK096467.1 - 4 498
AK225284.1 - 7 686
AK303611.1 - 8 917
AK307586.1 - 8 917
AY460334.1 - 8 884
BC011764.2 - 8 913 3616898
BC014559.2 - 8 913 4025673
BC016144.1 - 8 912
BC027252.1 - 7 609
BC033093.1 - 8 913 4577077
BX537490.1 - 4 1649
U40847.1 - 7 746
U76366.1 - 7 728

View Full Product Details