Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABC1
Entrez Gene ID
828127
Gene Name
ABC transporter 1
Gene Aliases
ABC transporter 1, AT4G01660, ATABC1, ATATH10, T15B16.14, T15B16_14
Chromosome Location
Chr.4: 708357 - 711256 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116396.4 NP_192075.1 3 - 4 1219 131

View Full Product Details