Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PGSIP4
- Entrez Gene ID
-
841934
- Gene Name
- plant glycogenin-like starch initiation protein 4
- Gene Aliases
-
AT1G54940, F14C21.47, F14C21_47, GUX4, Glucuronic Acid Substitution of Xylan 4, plant glycogenin-like starch initiation protein 4
- Chromosome Location
-
Chr.1: 20481648 - 20484799 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104367.2 |
NP_175891.1 |
2 - 3 |
1234 |
110 |
GenBank mRNA |
KJ138962.1 |
- |
2 - 3 |
1192 |
110 |
View Full Product Details